missing translation for 'onlineSavingsMsg'
Learn More

Streptomycin sulfate, MedChemExpress

MedChemExpress Streptomycin sulfate is an aminoglycoside antibiotic, that inhibits protein synthesis.

Encompass_Preferred

91.80 € - 122.85 €

Specifications

Chemical Name or Material Streptomycin sulfate
CAS 3810-74-0
Color White
Purity Grade Notes Research
Molecular Formula C21H42N7O18S1.5
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16488498
Encompass_Preferred
View Documents
Medchem Express
HY-B0472
10 g
91.80 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16404377
Encompass_Preferred
View Documents
Medchem Express
HY-B0472
50 g
122.85 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Streptomycin sulfate is an aminoglycoside antibiotic, that inhibits protein synthesis.
Specifications

Specifications

Streptomycin sulfate
White
C21H42N7O18S1.5
O[C@@]1(C=O)[C@H]([C@@H](O[C@H]1C)O[C@]([C@@H]([C@@H](O)[C@@H]2NC(N)=N)O)([H])[C@H]([C@@H]2O)NC(N)=N)O[C@@](O[C@@H](CO)[C@H](O)[C@H]3O)([H])[C@H]3NC.[1.5H2SO4]
728.69
Research
COVID-19-immunoregulation
3810-74-0
Research
H2O : ≥ 100 mg/mL (137.23 mM) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
728.69
98.0%
Solid
Videos
Safety and Handling

Safety and Handling

H302∣H361

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)

Recommended Storage : 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Streptomycin sulfate, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.