Learn More
Streptomycin sulfate, MedChemExpress
MedChemExpress Streptomycin sulfate is an aminoglycoside antibiotic, that inhibits protein synthesis.
Brand: Medchem Express HY-B0472

Description
Streptomycin sulfate is an aminoglycoside antibiotic, that inhibits protein synthesis.Specifications
Streptomycin sulfate | |
White | |
C21H42N7O18S1.5 | |
H2O : ≥ 100 mg/mL (137.23 mM) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) | |
728.69 | |
98.0% | |
Solid |
3810-74-0 | |
Research | |
50 g | |
O[C@@]1(C=O)[C@H]([C@@H](O[C@H]1C)O[C@]([C@@H]([C@@H](O)[C@@H]2NC(N)=N)O)([H])[C@H]([C@@H]2O)NC(N)=N)O[C@@](O[C@@H](CO)[C@H](O)[C@H]3O)([H])[C@H]3NC.[1.5H2SO4] | |
728.69 | |
Research | |
COVID-19-immunoregulation |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only