Learn More
Streptozocin, Tocris Bioscience™
DNA alkylator; antitumor and induces diabetes
Brand:  Tocris 1621/100
84.66 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18762714
  • 110.00 € / 100mg
Explore more special offers
Description
Description
Antibiotic and antitumor agent. Alkylates DNA and induces diabetes mellitus via reduction of nicotinamide adenine dinucleotide in pancreatic β-cells in vivo.
Specifications
Specifications
Streptozocin
100 mg
>95%
18883-66-4
C8H15N3O7