missing translation for 'onlineSavingsMsg'
Learn More

Strontium Titanium Oxide Substrate, 6.25 x 6.25 x 0.5mm, Polished One Side, 110 Orientation, Thermo Scientific™

100 Orientation

222.00 € - 752.00 €

Chemical Identifiers

CAS 12060-59-2
Molecular Formula O3SrTi
Molecular Weight (g/mol) 183.48
MDL Number MFCD00049554
InChI Key VEALVRVVWBQVSL-UHFFFAOYSA-N
Synonym strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
PubChem CID 82899
IUPAC Name strontium;dioxido(oxo)titanium
SMILES [Sr++].[O-][Ti]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11389228
SDS View Documents
Thermo Scientific Alfa Aesar
042617.KT
1 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11399228
SDS View Documents
Thermo Scientific Alfa Aesar
042617.KU
5 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

12060-59-2
183.48
VEALVRVVWBQVSL-UHFFFAOYSA-N
82899
[Sr++].[O-][Ti]([O-])=O
O3SrTi
MFCD00049554
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
strontium;dioxido(oxo)titanium

Specifications

12060-59-2
Substrate
MFCD00049554
VEALVRVVWBQVSL-UHFFFAOYSA-N
strontium;dioxido(oxo)titanium
82899
Strontium Titanium Oxide Substrate, Polished One Side
White
O3SrTi
strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
[Sr++].[O-][Ti]([O-])=O
183.48
Odorless
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 235-044-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.