missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium zirconium oxide, 99.3% (metals basis), Thermo Scientific Chemicals
50.30 € - 178.00 €
Chemical Identifiers
CAS | 12036-39-4 |
---|---|
Molecular Formula | O3SrZr |
Molecular Weight (g/mol) | 226.84 |
MDL Number | MFCD00049555 |
InChI Key | FCCTVDGKMTZSPU-UHFFFAOYSA-N |
Synonym | strontium zirconate, strontium zirconium trioxide, strontium dioxido oxo zirconium, o3zr.sr, strontium zirconium oxide srzro3, strontium zirconium oxide, strontium 2+ ion oxozirconiumbis olate, strontium 2+ oxozirconiumbis olate |
PubChem CID | 82852 |
IUPAC Name | strontium;dioxido(oxo)zirconium |
SMILES | [Sr++].[O-][Zr]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11321719
|
Thermo Scientific Alfa Aesar
012346.30 |
250 g |
50.30 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311719
|
Thermo Scientific Alfa Aesar
012346.A1 |
1 kg |
178.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mostly as intermediates( primary and secondary ). For chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12036-39-4 | |
226.84 | |
FCCTVDGKMTZSPU-UHFFFAOYSA-N | |
82852 | |
[Sr++].[O-][Zr]([O-])=O |
O3SrZr | |
MFCD00049555 | |
strontium zirconate, strontium zirconium trioxide, strontium dioxido oxo zirconium, o3zr.sr, strontium zirconium oxide srzro3, strontium zirconium oxide, strontium 2+ ion oxozirconiumbis olate, strontium 2+ oxozirconiumbis olate | |
strontium;dioxido(oxo)zirconium |
Specifications
9000°C | |
12036-39-4 | |
Powder | |
MFCD00049555 | |
FCCTVDGKMTZSPU-UHFFFAOYSA-N | |
strontium;dioxido(oxo)zirconium | |
82852 | |
99.3% | |
5.5 g/mL | |
Strontium zirconium oxide |
2700°C | |
White | |
O3SrZr | |
strontium zirconate, strontium zirconium trioxide, strontium dioxido oxo zirconium, o3zr.sr, strontium zirconium oxide srzro3, strontium zirconium oxide, strontium 2+ ion oxozirconiumbis olate, strontium 2+ oxozirconiumbis olate | |
[Sr++].[O-][Zr]([O-])=O | |
226.84 | |
226.84 | |
Odorless | |
(metals basis) |
Safety and Handling
missing translation for 'einecsNumber' : 234-847-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only