missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ HI-200 Series Hybridization Incubator Accessory, Tube
This glass HI-200 Series Hybridization Incubator Tube features a low probe volume, even with large glass tubes. The rotation and design ensures volumes as low as 5 mL can be used and recovered.
215.00 € - 245.00 €
Product Code | Brand | Length (Metric) | DiameterMetricInner | Closure Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | DiameterMetricInner | Closure Type | Price | Quantity & Availability | ||||
12142080
|
Cole-Parmer™
39455-17 |
205 mm | Screw-on End Caps |
245.00 €
Pack of 4 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
12093765
|
Cole-Parmer™
39455-10 |
200 mm | Not Included |
215.00 €
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
|||||
Description
- Tubes and other accessories can be accomodated at the same time for multiple uses.
- Unique slot-in tubes; with tube rotation speed of 0 to 20 rpm, controllable to suit your application.
- Radioactive safe; protective casing and non-drip tube design minimizes risk to user.
- Manufactured from thick, durable borosilicate glass.
- Easy pour - non-drip, safe, and convenient.
- Sealing ring - O-ring sealed end caps ensure no leakage.
- Escape thread on screw cap - easy opening with no vacuum problems.
Specifications
Borosilicate Glass | |
Hybridization Tube |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ HI-200 Series Hybridization Incubator Accessory, Tube