missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Tube Holder

Tube Holder for Use with Microtitre Plate Shakers SSM5 and SSL5.

Brand:  Cole-Parmer™ 51900-98

 View more versions of this product

Product Code. 12944025

  • 172.00 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Accessory tube holders accommodate 0.2 mL or 0.5 mL microcentrifuge tubes.
  • Two tube holders can be used at any one time and securely held in place by the non-slip mat.
  • Stuart™ Tube Holder; For Use with Microtitre Plate Shakers where samples require controlled temperature and/or humidity conditions, ideal for placement inside CO2incubators.
Specifications

Specifications

0.5 mL Tubes
Tube Holder
1 Ea.
0.5 mL
Stuart™ Microtitre Plate Shakers SSM5 and SSL5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.