Learn More
Cole-Parmer™ Tube Holder
Tube Holder for Use with Microtitre Plate Shakers SSM5 and SSL5.
165.00 € - 238.00 €
Specifications
Product Type Tube Holder
For Use With (Equipment) Stuart™ Microtitre Plate Shakers SSM5 and SSL5
Packaging Quantity 1 Ea.
View More Specs
Products 3
Product Code Brand Holds Price Quantity & Availability  
Product Code Brand Holds Price Quantity & Availability  
12693335
View Documents
Cole-Parmer™
51900-99
0.2 mLTubes
165.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
12944025
View Documents
Cole-Parmer™
51900-98
0.5 mL Tubes
172.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
11496608
View Documents
Cole-Parmer™
51900-97
1.5 mL Tubes
238.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
Description
Description
  • Accessory tube holders accommodate 0.2 mL or 0.5 mL microcentrifuge tubes.
  • Two tube holders can be used at any one time and securely held in place by the non-slip mat.
  • Stuart™ Tube Holder; For Use with Microtitre Plate Shakers where samples require controlled temperature and/or humidity conditions, ideal for placement inside CO2incubators.
Specifications
Specifications
Tube Holder
Stuart™ Microtitre Plate Shakers SSM5 and SSL5
1 Ea.