missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic acid, ≥99.5% (alkalimetric), Honeywell™

≥99.5% (alkalimetric)

51.46 € - 505.00 €

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.088
MDL Number MFCD00011603
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15660860
View Documents
Honeywell Research Chemicals
07401-1KG
1 kg
51.46 €
1kg
Please to purchase this item. Need a web account? Register with us today!
15670860
View Documents
Honeywell Research Chemicals
07401-2.5KG
2.5 kg
101.47 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
15600870
View Documents
Honeywell Research Chemicals
07401-5KG
5 kg
187.61 €
5kg
Please to purchase this item. Need a web account? Register with us today!
15610870
View Documents
Honeywell Research Chemicals
07401-6X2.5KG
6 x 2.5 kg
505.00 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15690860
View Documents
Honeywell Research Chemicals
07401-4X5KG
4 x 5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Chemical Identifiers

5329-14-6
97.088
IIACRCGMVDHOTQ-UHFFFAOYSA-N
5987
sulfamic acid
H3NO3S
MFCD00011603
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
CHEBI:9330
NS(=O)(=O)O

Specifications

5329-14-6
H3NO3S
MFCD00011603
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
NS(=O)(=O)O
97.088
CHEBI:9330
≥99.5% (alkalimetric)
≤0.02%
NH2SO3H
2967
IIACRCGMVDHOTQ-UHFFFAOYSA-N
sulfamic acid
5987
97.09g/mol
Sulfamic Acid
Videos
Safety and Handling

Safety and Handling

P273-P305 + P351 + P338

H315-H319-H412

missing translation for 'einecsNumber' : 226-218-8

Product Identifier
  • Sulfamic acid

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : sulphamidic acid

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.