missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SulfoLink™ Coupling Resin

Covalently immobilize cysteine-peptides or proteins (sulfhydryl-ligands) for affinity purification using iodoacetyl-activated agarose beads and columns.

Brand:  Thermo Scientific™ 20404

2926.00 EUR valid until 2025-12-31
Use promo code "24353" to get your promotional price.


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11796406

  • 3240.00 € / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Upon receipt store product in the dark at 4°C. Product is shipped at ambient temperature.
250 mL
Haloacetyl
Bottle
SulfoLink™
Resin
1 mg Peptide or 10 mg Antibody per mL of Resin
Affinity, Agarose Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL405926 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number20404
Lot NumberZI402986 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number20404
Lot NumberZE383012 Certificate TypeCertificate of Analysis Date22/05/2024 Catalog Number20404
Lot NumberSPECS Certificate TypeCertificate of Analysis Date22/05/2024 Catalog Number20404
Lot NumberYK371381 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog Number20404
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ SulfoLink™ Coupling Resin > SulfoLink Coupling Resin; 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.