Learn More
Sulfur Certified Reference Material, UKAS ISO 17025 & ISO 17034 Certified, Sulfur in Synthetic Diesel, 0.0010 % (10ppm), 100mL., Paragon Scientific™
Paragon Scientific’s Synthetic Diesel Sulfur CRMs are suitable for calibration, verification or for use in quality control procedures used for analysing sulfur content in petroleum products, with sulfur content ranging from 0 (Blank) through to 5.0000% (50000ppm) for each material.
Brand:  Paragon Scientific GOSU-02
Product Code. 16040150
  • 117.00 € / 100mL
Explore more special offers
Description
Description
  • Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation.
  • Highest level of accreditation guarantee, providing the most credible certified data available worldwide.
  • Suitable for but not limited to internationally recognised test methods and protocol: ASTM D2622, ASTM D4294, ASTM D5453, ASTM D7039, IP 336, IP 496, IP 497.
  • Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data.
  • Supplied in 100 mL volume, tamper-evident glass packaging, providing assurance of sample integrity.
  • Manufactured in the United Kingdom.
  • 12 month shelf life.
Specifications
Specifications
100 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sulfur Certified Reference Material, UKAS ISO 17025 & ISO 17034 Certified, Sulfur in Synthetic Diesel, 0.0010 % (10ppm), 100mL., Paragon Scientific™ > Quantity: 100 mL
Nominal values quoted. Certified Values will vary from batch to batch.