missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Min 90% d=1.83, SLR, Extra Pure, for Milk Testing, (Gerber Test), Fisher Chemical™
H2O4S, CAS Number-7664-93-9, electrolyte acid, dipping acid, dihydrogen sulfate, acide sulfurique, acidum sulfuricum, battery acid, sulphuric acid, mattling acid, oil of vitriol, vitriol brown oil, 2.5 L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid
50.60 € - 94.25 €
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15938916
|
Fisher Chemical
S/9360/PB15 |
1 L | HDPE Plastic Bottle |
50.60 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10427080
|
Fisher Chemical
S/9360/PB17 |
2.5 L | HDPE Plastic Bottle |
94.25 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
H2O4S, CAS Number-7664-93-9, electrolyte acid, dipping acid, dihydrogen sulfate, acide sulfurique, acidum sulfuricum, battery acid, sulphuric acid, mattling acid, oil of vitriol, vitriol brown oil, 2.5 L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
CHEBI:26836 | |
OS(O)(=O)=O |
H2O4S | |
MFCD00064589 | |
1118 | |
sulfuric acid |
Specifications
1 | |
7664-93-9 | |
290°C | |
H2O4S | |
Liquid | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 |
Colorless | |
10°C | |
1mmHg 146 | |
21mPa at 25°C | |
MFCD00064589 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.07g/mol |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.