missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid solution, 0.1M, Reag. Ph. Eur., Honeywell Fluka™

Volumetric, Reag. Ph. Eur., 0.1 M H2SO4 (0.2N)

77.05 € - 449.82 €

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15694990
View Documents Promotion Details
Honeywell-Fluka
35357-1L
1 L
77.05 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15615000
View Documents Promotion Details
Honeywell-Fluka
35357-5L-VP
5 L
258.77 €
5 L
Please to purchase this item. Need a web account? Register with us today!
15605000
View Documents Promotion Details
Honeywell Fluka
35357-5L
5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15625000
View Documents Promotion Details
Honeywell-Fluka
35357-6X1L
6 x 1 L
401.94 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15684990
View Documents Promotion Details
Honeywell-Fluka
35357-10L-VP
10 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15674990
View Documents Promotion Details
Honeywell Fluka
35357-10L
10 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O

Specifications

7664-93-9
H2O4S
MFCD00064589
2037554
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08g/mol
Sulfuric acid solution
0.1M
H2SO4
UN3264
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
Reagent Ph. Eur.
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sulfuric acid solution

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.

Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid

SDS
Documents

Documents

Product Certifications