Learn More
Epredia™ SuperFrost™ Microscope Slides
The printed Epredia™ SuperFrost tab is impervious to virtually all common laboratory chemicals and reagents. A wide range of colors allows several examination specimens to be labeled and archived simply and clearly.
Brand:  Epredia™ AG00008132E01MNZ20
 View more versions of this product
Product Code. 17294884
  • 7.15 € / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Time-consuming labeling with paper labels is no longer required
  • The writing area is resistant to all common solvents
  • SuperFrost slides may be sorted by color e.g. by patient, urgency, stain or examination method
  • Simple recognition of specified characteristics such as year of examination or department, use etc.
Specifications
Specifications
90°, Ground
Blue
Uncoated
76 x 26 mm
1 mm
ISO8037/I
Yes