missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ SuperFrost™ Microscope Slides
The printed Epredia™ SuperFrost tab is impervious to virtually all common laboratory chemicals and reagents. A wide range of colors allows several examination specimens to be labeled and archived simply and clearly.
6.80 € - 7.15 €
Specifications
Edges | 90°, Ground |
---|---|
Thickness (Metric) | 1 mm |
Certifications/Compliance | ISO8037/I |
Coating | Uncoated |
Writing Surface | Yes |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
17294884
|
Epredia™
AG00008132E01MNZ20 |
Blue |
7.15 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
17204894
|
Epredia™
AG00008232E01MNZ20 |
Pink |
7.15 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
17214894
|
Epredia™
AG00008332E01MNZ20 |
Yellow |
6.80 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
17224894
|
Epredia™
AG00008432E01MNZ20 |
Green |
6.80 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
Description
- Time-consuming labeling with paper labels is no longer required
- The writing area is resistant to all common solvents
- SuperFrost slides may be sorted by color e.g. by patient, urgency, stain or examination method
- Simple recognition of specified characteristics such as year of examination or department, use etc.
Specifications
90°, Ground | |
ISO8037/I | |
Yes |
1 mm | |
Uncoated | |
76 x 26 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ SuperFrost™ Microscope Slides