Learn More
Cytiva Superose™ 6 Increase Columns
High-resolution size-exclusion chromatography for studying large proteins and complexes. Cytiva Lifescience™ Superose™ 6 Increase Columns delivers high quality in size exclusion chromatography (SEC).
Brand:  Cytiva 29091597
 View more versions of this product
Product Code. 15383224
  • 2865.00 € / Each
Explore more special offers
Description
Description
Superose™ 6 Increase pre-packed columns deliver the quality and high resolution in size exclusion chromatography (SEC) — even for large proteins and aggregates.
Increased performance
  • Higher resolution than traditional Superose 6 columns
  • Wide fractionation range (Mr from 5,000 to 5,000,000)
  • Withstands high-pH cleaning
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Superose 6 Increase 5/150 GL
150 mm
Borosilicate glass
435 psi
8.6μm
4° to 30° in 20% ethanol
5 mm
0.3mL/min.
Cross-linked agarose
3 to 12
Polyethylene (PE) filter
Gel Filtration Column
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Superose™ 6 Increase Columns > Superose 6 Increase 5/150 GL