Learn More
Invitrogen™ SureCast™ Gel Handcast Bundle B - Hardware Only
Pour your own polyacrylamide gels easily and confidently with a SureCast™ Gel Handcast Bundle B. The SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.
Brand:  Invitrogen™ HC1000S
241.56 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Product Code. 15597731
  • 409.00 € / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Includes:
The SureCast Handcast Bundle B includes all the hardware you need to get started:
  • 2 SureCast Handcast Stations
  • 2 SureCast™ Sealing Pads
  • 2 SureCast™ Glass Plate Sets (2 front and 2 back)
  • 6 SureCast™ Gel Combs (2 each: 10-, 12-, 15-well)
  • 10 SureCast™ Gel Spacers
Description
Description
  • Innovative, leak-free design—minimizes the number of failed gels, resulting in less wasted time
  • Superior glass plate durability
  • Simple assembly of casting components—just drop the glass plates in, then lock the cam handle
  • Unique tilt feature for easy loading of acrylamide solutions
The complete product includes the SureCast™ Handcast System. For a bundle that also includes the SureCast™ Handcast Reagents, please see SureCast™ Gel Handcast Bundle A. The system features durable glass plates that are securely sealed using a single-motion, load-and-lock mechanism. The unique tilt feature of the SureCast™ Handcast Station minimizes spillage and enhances loadability of casting solutions during hand casting. SureCast gels are designed for use in the Mini Gel Tank.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Manuals
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ SureCast™ Gel Handcast Bundle B - Hardware Only > Casting equipment and accessories-only bundle