Learn More
Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips
GREENER_CHOICE
Choose from a comprehensive line of pipet tips for demanding liquid-handling applications
Brand:  Fisherbrand™ TF140-300-FIS
47.59 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
 View more versions of this product
Product Code. 11987714
  • 135.00 € / Pack of 960
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Choose from a comprehensive line of pipet tips for demanding liquid-handling applications
Universal fit for research grade pipetters.
  • Engineered for increased accuracy and are compatible with single-channel, multichannel, and electronic pipetters
  • DNase-/RNase-free; sterile tips are also pyrogen-free
  • Sterile and clear color
  • Filter eliminates aerosol cross-contamination without sealing or trapping samples
  • Tips allow for noncontaminated sample recovery
  • Not autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
30 to 300 μL
Sterile
Not Autoclavable
58 mm
Micro Point
Yes
10 μL, 50 μL, 100 μL, 200 μL
Filtered
Clear
2 in.
Hinged Rack
Universal Fit
SureOne
Product Suggestions
Product Suggestions
Videos
Videos
  • Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips - POTM - April (EN)
SDS
Documents Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips > 30 to 300μL; Micropoint tip; Graduated at 10, 50, 100, 200Glc muL