missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A&D Instruments™ SV-10 Series Sine Wave Vibro Viscometer

The SV-10 tuning fork vibration viscometer is simple to use and provides highly accurate, fast results. The instrument is ideal for Newtonian and Non-Newtonian samples, especially those of a volatile nature.
Brand: A&D Instruments™ SV-10
This item is not returnable.
View return policy
Description
- Measurement of very low viscosities (down to 1/3 of water viscosity)
- Small sample size, standard sample cups 35 mL with optional size down to 13 mL and 10 mL
- No spindles or interchangeable parts
- Easy to clean, just wipe down
- Standard temperature measurement from 0°C to 160°C
- First result within 15 seconds, then continuous real-time data
- Use standalone or with Free PC software for data collection and graphing in real-time
- Wide measurement range: 0.3 to 10,000 mPa (SV-10), 1000 to 100,000 mPa (SV-100)
- High precision with a repeatability of ±1% of displayed value
Specifications
0.3 to 10,000 cP | |
RS232C Interface, 1.5 m Connection Cable | |
332 mm | |
0°C to 160°C | |
AC Adapter, 14 VA |
VFD | |
536 mm | |
314 mm | |
5 kg | |
30 Hz |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
A&D Instruments™ SV-10 Series Sine Wave Vibro Viscometer > Sine Wave Vibro Viscometer; Depth (Metric) Bath: 314 mm; Display Type: VFD; Temperature Range (Metric): 0°C to 160°C; Viscosity Range: 0.3 to 10,000 cP; Includes: RS232C Interface, 1.5 m Connection Cable
Spot an opportunity for improvement?Share a Content Correction