missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SVL™ Torion Spare Fitting Joints
For use with Torion glass bodies and rubber/PTFE sealing ring
Brand: SVL 720-06
This item is not returnable.
View return policy
Description
- Glass filled polypropylene for use at temperatures up to 180°C
- PTFE for use at temperatures up to 250°C
Specifications
10/Pk. | |
Torion spares | |
PTFE clamp | |
14.7mm (Bore) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction