missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SVL™ Torion Spare Fitting Joints
For use with Torion glass bodies and rubber/PTFE sealing ring
90.05 € - 1650.00 €
Product Code | Brand | Thickness (Metric) Wall | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thickness (Metric) Wall | Type | Price | Quantity & Availability | ||||
12698405
|
SVL
720-05 |
8.7mm (Bore) | PTFE Clamp | N/A | |||||
12563346
|
SVL
720-03 |
14.7mm (Bore) | Polypropylene Clamp |
120.00 €
Pack of 10 |
Estimated Shipment: 10-03-2025
Log in to see stock. |
||||
15312969
|
SVL
720-01 |
5.7mm (Bore) | Polypropylene clamp |
90.05 €
Pack of 10 |
Estimated Shipment: 10-03-2025
Log in to see stock. |
||||
12493531
|
SVL
720-06 |
14.7mm (Bore) | PTFE clamp |
1650.00 €
Pack of 10 |
Estimated Shipment: 10-03-2025
Log in to see stock. |
||||
13431819
|
SVL
720-02 |
8.7mm (Bore) | Polypropylene Clamp |
94.80 €
Pack of 10 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||||
Description
- Glass filled polypropylene for use at temperatures up to 180°C
- PTFE for use at temperatures up to 250°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title