missing translation for 'onlineSavingsMsg'
Learn More

SVL™ Torion Spare Fitting Joints

For use with Torion glass bodies and rubber/PTFE sealing ring

Brand:  SVL 720-05

 View more versions of this product

Product Code. 12698405

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Glass filled polypropylene for use at temperatures up to 180°C
  • PTFE for use at temperatures up to 250°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTFE Clamp
8.7mm (Bore)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers