Learn More
Tuttnauer Switch Box
Switch Box from 3-Phases to a single phase. The current electricity for machine with 9 kW steam generator is 40 A.
Brand:  Tuttnauer ELC258-0001
Product Code. 17849548
  • 269.00 € / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Switch box from 3-Phases to a single phase.
  • The current electricity for machine with 9 kW steam generator is 40 A.
Specifications
Specifications
Switch Box
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tuttnauer Switch Box > Switch Box