missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Tacta™ Mechanical Pipet Multipacks
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Multipacks combine a set of Tacta single-channel mechanical pipets with matching tip trays and useful accessories. Sartorius Tacta Mechanical Pipet Multipacks provides best alternative to replace your used mechanical pipets.
Brand: Sartorius LH-729670
867.99 EUR valid until 2025-12-31
Use promo code "24294" to get your promotional price.
Includes: 3 x Single-channel pipets (0.5 to 10μL, 10 to 100μL, 100 to 1000μL), Tip Racks of 96 tips (0.2 to 10μL, 0.5 to 200μL, 10 to 1000μL), 1 x Linear Stand, 3 x Single Pipet Holders
Description
- Comfortable to hold due to ergonomically designed handle
- Low pipetting and tip ejection forces
- Smooth tip ejection with the new Optiject™ technology
- Unique Optilock™ system provides flexibility for volume adjustment and locking
- Calibration adjustment to provide accurate results for various liquid types
- Reliable results, even over lengthy pipetting periods
- Easy-to-read, four-digit volume display helps to set exact volumes
- Easy to clean with only three parts to disassemble
- Fully autoclavable
- High-quality standard Optifit tips for various applications
Services
Two years
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Autoclavable | |
1 | |
Mechanical Pipette Multipacks | |
Four-digit volume display | |
10/Pk. | |
2 Years | |
79 g (1-channel, 1000μL); 138 g (8-channel, 300μL) |
Tacta | |
Tacta | |
0.5-10 μL, 10-100 μL, 100-1000 μL | |
3 x Single-channel pipets (0.5 to 10μL, 10 to 100μL, 100 to 1000μL), Tip Racks of 96 tips (0.2 to 10μL, 0.5 to 200μL, 10 to 1000μL), 1 x Linear Stand, 3 x Single Pipet Holders | |
High | |
138 mm (1-channel, 1000μL); 240 mm (8-channel, 300μL) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction