missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Tamper-Evident Screw Cap
Tamper-Evident Screw Cap equipped with a silicone seal is suitable for use with microtubes.
Brand: BRAND™ 780772
147.62 EUR valid until 2024-10-31
Use promo code "21661" to get your promotional price.
This item is not returnable.
View return policy
Description
- With silicone seal
- For microtubes
- Working temperature between -196 to 121°C
Specifications
Silicone Seal | |
Tamper-evident Screw Cap | |
Green |
Silicone | |
Polypropylene | |
78076064 Microtubes |