missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tanshinone I, MedChemExpress
MedChemExpress Tanshinone I is an inhibitor of type IIA human recombinant sPLA2 (IC50=11 μM) and rabbit recombinant cPLA2 (IC50=82 μM).
66.15 € - 359.10 €
Specifications
Chemical Name or Material | Tanshinone I |
---|---|
CAS | 568-73-0 |
Color | Dark Brown |
Purity Grade Notes | Research |
Molecular Formula | C18H12O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16483836
|
Medchem Express
HY-N0134 |
25 mg |
186.30 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16468996
|
Medchem Express
HY-N0134 |
100 mg |
359.10 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16434767
|
Medchem Express
HY-N0134 |
50 mg |
252.45 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16418878
|
Medchem Express
HY-N0134 |
10 mg |
93.15 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16411318
|
Medchem Express
HY-N0134 |
5 mg |
66.15 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tanshinone I is an inhibitor of type IIA human recombinant sPLA2 (IC50=11 μM) and rabbit recombinant cPLA2 (IC50=82 μM).Specifications
Tanshinone I | |
Dark Brown | |
C18H12O3 | |
DMSO : 2 mg/mL (7.24 mM; Need ultrasonic) | |
276.29 | |
98.0% | |
Solid |
568-73-0 | |
Research | |
Tanshinone A | |
O=C(C1=C2C=CC3=C1C=CC=C3C)C(C4=C2OC=C4C)=O | |
276.29 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only