Learn More
Tanshinone IIA sulfonate sodium, MedChemExpress
MedChemExpress Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA, which acts as an inhibitor of store-operated Ca2+ entry (SOCE), and is used to treat cardiovascular disorders.
Brand: Medchem Express HY-N1370
This item is not returnable.
View return policy
Description
Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA, which acts as an inhibitor of store-operated Ca2+ entry (SOCE), and is used to treat cardiovascular disorders.Specifications
Tanshinone IIA sulfonate sodium | |
wine | |
C19H17NaO6S | |
Sodium Tanshinone IIA sulfonate Tanshinone IIA sodium sulfonate | |
O=S(C1=C(C)C(C(C(C2=C3C=CC4=C2CCCC4(C)C)=O)=O)=C3O1)(O[Na])=O | |
396.39 | |
Research | |
COVID-19-immunoregulation |
69659-80-9 | |
Research | |
10 mg | |
DMSO : 62.5 mg/mL (157.67 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (21.01 mM; Need ultrasonic) | |
396.39 | |
98.0% | |
Solid |
Research purposes only