missing translation for 'onlineSavingsMsg'
Learn More

Tanshinone IIA sulfonate sodium, MedChemExpress

MedChemExpress Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA, which acts as an inhibitor of store-operated Ca2+ entry (SOCE), and is used to treat cardiovascular disorders.

66.15 € - 106.65 €

Specifications

Chemical Name or Material Tanshinone IIA sulfonate sodium
CAS 69659-80-9
Color wine
Purity Grade Notes Research
Molecular Formula C19H17NaO6S
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16453916
Encompass_Preferred
View Documents
Medchem Express
HY-N1370
25 mg
106.65 €
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16409218
Encompass_Preferred
View Documents
Medchem Express
HY-N1370
10 mg
66.15 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA, which acts as an inhibitor of store-operated Ca2+ entry (SOCE), and is used to treat cardiovascular disorders.
Specifications

Specifications

Tanshinone IIA sulfonate sodium
wine
C19H17NaO6S
DMSO : 62.5 mg/mL (157.67 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (21.01 mM; Need ultrasonic)
396.39
98.0%
Solid
69659-80-9
Research
Sodium Tanshinone IIA sulfonate Tanshinone IIA sodium sulfonate
O=S(C1=C(C)C(C(C(C2=C3C=CC4=C2CCCC4(C)C)=O)=O)=C3O1)(O[Na])=O
396.39
Research
COVID-19-immunoregulation
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣The compound is unstable in solutions, freshly prepared is recommended.

Recommended Storage : 4°C, sealed storage, away from moisture∣The compound is unstable in solutions, freshly prepared is recommended.

SDS
Documents

Documents

Product Certifications

Research purposes only