Learn More
Tantalum tris(diethylamido)-tert-butylimide, 99.99% (metals basis), Thermo Scientific Chemicals
Precursor for TaN
62.20 € - 908.00 €
Chemical Identifiers
CAS | 169896-41-7 |
---|---|
Molecular Formula | C15H39N4Ta |
Molecular Weight (g/mol) | 456.45 |
MDL Number | MFCD02684506 |
InChI Key | NXXDVZOFTYVIKX-UHFFFAOYSA-N |
Synonym | tert-butylimino tris diethylamino tantalum, tantalum tris diethylamido-tert-butylimide, tbtdet, tris diethylamino tert-butylimino tantalum, t-butylimido tris diethylamino tantalum v, tris diethylamido tert-butylimido tantalum v, tris diethylamino tert-butylimino tantalum v, tert-butylimino bis diethylamino tantalio diethylamine, tantalum,tris n-ethylethanaminato 2-methyl-2-propanaminato 2--, t-4 |
PubChem CID | 4100858 |
IUPAC Name | tert-butyliminotantalum;diethylazanide |
SMILES | CCN(CC)[Ta](N(CC)CC)(N(CC)CC)=[N](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11375707
|
Thermo Scientific Alfa Aesar
043419.03 |
1 g | |||||||
11365707
|
Thermo Scientific Alfa Aesar
043419.06 |
5 g |
242.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11355707
|
Thermo Scientific Alfa Aesar
043419.14 |
25 g |
908.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tantalum tris(diethylamido)-tert-butylimide is used as a precursor for tantalum nitride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
169896-41-7 | |
456.45 | |
NXXDVZOFTYVIKX-UHFFFAOYSA-N | |
4100858 | |
CCN(CC)[Ta](N(CC)CC)(N(CC)CC)=[N](C)(C)C |
C15H39N4Ta | |
MFCD02684506 | |
tert-butylimino tris diethylamino tantalum, tantalum tris diethylamido-tert-butylimide, tbtdet, tris diethylamino tert-butylimino tantalum, t-butylimido tris diethylamino tantalum v, tris diethylamido tert-butylimido tantalum v, tris diethylamino tert-butylimino tantalum v, tert-butylimino bis diethylamino tantalio diethylamine, tantalum,tris n-ethylethanaminato 2-methyl-2-propanaminato 2--, t-4 | |
tert-butyliminotantalum;diethylazanide |
Specifications
169896-41-7 | |
90°C (0.1 mmHg) | |
C15H39N4Ta | |
UN3129 | |
tert-butylimino tris diethylamino tantalum, tantalum tris diethylamido-tert-butylimide, tbtdet, tris diethylamino tert-butylimino tantalum, t-butylimido tris diethylamino tantalum v, tris diethylamido tert-butylimido tantalum v, tris diethylamino tert-butylimino tantalum v, tert-butylimino bis diethylamino tantalio diethylamine, tantalum,tris n-ethylethanaminato 2-methyl-2-propanaminato 2--, t-4 | |
NXXDVZOFTYVIKX-UHFFFAOYSA-N | |
tert-butyliminotantalum;diethylazanide | |
4100858 | |
99.99% | |
(metals basis) |
Yellow | |
Ampule | |
MFCD02684506 | |
Air and moisture sensitive | |
Reacts with water. | |
CCN(CC)[Ta](N(CC)CC)(N(CC)CC)=[N](C)(C)C | |
456.45 | |
468.48 | |
Liquid | |
Tantalum tris (diethylamido)-tert-butylimide |
Safety and Handling
GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H261-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
- Tantalum tris(diethylamido)-tert-butylimide
Signal Word
- Danger
Hazard Category
- Substance which\, in contact with water\, emits flammable gases Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H261-In contact with water releases flammable gases.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P231+P232-Handle under inert gas. Protect from moisture.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only