missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) oxide, 99.95% (metals basis excluding Nb), Nb <200ppm, Thermo Scientific Chemicals

179.00 € - 497.00 €

Chemical Identifiers

CAS 1314-61-0
Molecular Formula O5Ta2
Molecular Weight (g/mol) 441.89
MDL Number MFCD00011254
InChI Key PBCFLUZVCVVTBY-UHFFFAOYSA-N
IUPAC Name [(dioxotantalio)oxy]dioxotantalum
SMILES O=[Ta](=O)O[Ta](=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11333428
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036343.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11343428
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036343.22
100 g
497.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tantalum (V) oxide (Ta2O5) has potential applications as part of an environmental barrier coating system for Si3N4-based turbine components. Octadecyl phosphoric acid ester has been found to produce oriented, well-ordered monolayers on a flat tantalum (V) oxide surface, via self-assembly from a heptane/propan-2-ol solution. Tantalum(V) oxide (Ta 2 O 5 ) has been used extensively as an insulating layer in discrete capacitors and shows a high potential for semiconductor applications such as an insulating dielectric in the capacitive element in memory devices such as dynamic random access.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-61-0
441.89
PBCFLUZVCVVTBY-UHFFFAOYSA-N
O=[Ta](=O)O[Ta](=O)=O
O5Ta2
MFCD00011254
[(dioxotantalio)oxy]dioxotantalum

Specifications

1800°C
Sintered Lump
MFCD00011254
Insoluble in water.
O=[Ta](=O)O[Ta](=O)=O
441.89
99.95%
8.2 g/mL
Tantalum(V) oxide
1314-61-0
O5Ta2
14,9056
PBCFLUZVCVVTBY-UHFFFAOYSA-N
[(dioxotantalio)oxy]dioxotantalum
441.89
Odorless
(metals basis excluding Nb), Nb <200ppm
Videos
Safety and Handling

Safety and Handling

H500

missing translation for 'einecsNumber' : 215-238-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only