missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Spigot for Tank

Constructed of high density polyethylene, making it good for packaging and transporting liquids in the food industry. Kartell™ Spigot for Tank facilitates easy use by allowing the tank to be in a position to stand.

Brand:  Kartell™ 0177200

Product Code. 12801773

  • 12.25 € / Pack of 2
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with 10L and 20L tanks
  • Spigot is placed in a position allowing the tank to stand, as to facilitate the use of these tanks in packaging and transporting liquids
  • Suitable for foodstuff
Specifications

Specifications

36.50 mm
White
HDPE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kartell™ Spigot for Tank > ClosureSize: 36.50mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.