missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Taq DNA Polymerase, native (5 U/μL)

Thermo Scientific Native Taq DNA Polymerase is a highly thermostable DNA polymerase of the thermophilic bacterium Thermus aquaticus.

Brand:  Thermo Scientific™ EP0072

 View more versions of this product

Product Code. 10756421

  • 232.00 € / 500 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Taq DNA Polymerase, native, is a highly thermostable DNA polymerase of the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5' to 3' synthesis of DNA, has no detectable 3' to 5' exonuclease (proofreading) activity, and possesses low 5' to 3' exonuclease activity. In addition, the polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Native Taq DNA Polymerase is preferred for amplification of bacterial DNA sequences homologous to those found in E. coli.

Available with or without the stabilizing agent BSA, Taq DNA Polymerase, native, with BSA, is often considered the best choice when amplifying DNA samples of lower purity, e.g., genomic DNA from mouse tail.

Both native Taq DNA polymerases are supplied with two buffers: 10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4. The latter allows for PCR at a wide range of magnesium concentrations and decreases unspecific priming

Features

  • Thermostable with half-life more than 40 min at 95°C
  • Generates PCR products with 3'-dA overhangs
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescent-labeled nucleotides)

Applications

  • Routine PCR amplification of DNA fragments up to 5 kb
  • DNA labeling
  • High throughput PCR

Notes

  • The error rate of Taq DNA Polymerase in PCR is 2.2 x 10-5 errors per nt per cycle. Accordingly, the accuracy of PCR is 4.5 x 104. Accuracy is an inverse of the error rate and shows an average number of correct nucleotides incorporated before an error occurs.
  • The 10X Taq Buffer without Detergent is recommended for microarray experiments.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Taq DNA Polymerase, native (5 U/μL), 100 μL
• 10X Taq Buffer with KCl, 2 x 1.25 mL
• 10X Taq Buffer with (NH4)2SO4, 2 x 1.25 mL
• 25 mM MgCl2, 2 x 1.25 mL
• BSA (20 mg/mL), 0.25 mL

Store at -20°C.
Taq DNA Polymerase
DNA Polymerase
PCR
Standard PCR
With BSA
72°C
500 units
Dry Ice
5 U/μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.