missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ β-Actin Detection Reagents
TaqMan™ β-Actin Detection Reagents
Brand: Applied Biosystems™ 401846
This item is not returnable.
View return policy
Description
Applied Biosystems™ TaqMan™ β-actin provides the necessary components for using β-actin as a normalization control in TaqMan™ real-time PCR applications employing human nucleic acid. Either DNA or RNA can be used as a target template.Notes on Use
The TaqMan™ β-actin Detection Reagent kit can be used with TaqMan™ One-Step RT-PCR Master Mix Reagents, TaqMan™ EZ RT-PCR Core Reagents, or the TaqMan™ Gold RT-PCR Reagents for one-step reverse transcription polymerase chain reaction (RT-PCR).
Notice to Purchaser
1. Disclaimer of License: These products may be useful in the Polymerase Chain Reaction (PCR) methods covered by patents owned by Roche Molecular Systems, Inc. and F. Hoffmann-La Roche Ltd. However, no license under these patents to use the PCR process is conveyed expressly or by implication to the purchaser by the purchase of this product.
TaqMan™ probes are covered by US Patent 5,723,591 and foreign counterparts and patents pending owned by PE Corporation (NY), and may be covered by US Patents 5,801,155 and 6,084,102 and foreign counterparts licensed to Applied Biosystems™
Note: See user's manual or package insert for limited label license, and trademark information. For Research Use Only. Not for use in diagnostics procedures.

Specifications
Control Template(s) | |
Tube | |
ACTB | |
FAM | |
Real Time PCR Reagents | |
Dry Ice | |
100 Reactions | |
100 Reactions |
Human Control DNA | |
1-step RT-qPCR | |
TAMRA™ Quencher (3'), FAM (5') | |
TaqMan™ | |
100 reactions | |
Human | |
β-actin |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Protocol: TaqMan® PCR Reagent Kit with Controls: With Amplitaq Gold® DNA Polymerase (English )
Version: 19 Aug 2010
File size: 1.1 MB
Protocol: TaqMan® Ribosomal RNA Control Reagents VIC™ Probe: Protocol: Rev C (English )
Version: 20 Jun 2007
File size: 637.4 kB
Essentials of Real Time PCR
File size: 153.6 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Applied Biosystems™ TaqMan™ β-Actin Detection Reagents > 100 reactions
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction