missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array, Human ABC Transporters, Fast 96-well
TaqMan™ Array, Human ABC Transporters, Fast 96-well
Brand: Applied Biosystems™ 4418811
This item is not returnable.
View return policy
Description
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan™ Array Human ABC Transporters 96-well Plate contains 44 assays to ABC transporter associated genes and 4 assays to candidate endogenous control genes. All assays are plated in duplicate.
Specifications
Array Plate | |
Human | |
Dried Down | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
Fast |
96-array Plate | |
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C. | |
TaqMan Assay for One 10 uL Reaction | |
FAM | |
1 x 96-well plate | |
Neurobiology, Biomarker-Related Pathway(s), Oncology, Signal Transduction |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Human ABC Transporters
Version: version 1.1
File size: 11.5 kB
Product Bulletin: TaqMan Array Plates
File size: 1 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Applied Biosystems™ TaqMan™ Array, Human ABC Transporters, Fast 96-well > 1 x 96 well plate
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction