missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TaqMan™ Fast Advanced Cells-to-CT™ Kit
GREENER_CHOICE

The TaqMan Fast Advanced Cells-to-Ct Kit is a complete cell lysate system designed for gene expression analysis directly from cultured cells without RNA purification.

Brand:  Invitrogen™ A35377

1655.80 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 16504654

  • 1948.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Cell lysis reagent, DNAse I, Stop Solution, 20X RT enzyme mix, 2X RT Buffer, TaqMan Fast Advanced Master Mix
This item is not returnable. View return policy

Description

Description

The TaqMan Fast Advanced Cells-to-Ct Kit is a complete cell lysate system designed for gene expression analysis directly from cultured cells without RNA purification. TaqMan Fast Advanced Cells-to-Ct cell lysis reagents are supplied with highest sensitivity Fast Advanced reverse transcription enzyme mix, 2X RT buffer, and best-in-class TaqMan Fast Advanced master mix. The TaqMan Fast Advanced Cells-to-Ct Kit gives you all the sensitivity you need in the fastest workflow available: get gene expressions analysis results from an entire plate of cells in about 80 minutes.

  • Complete—optimized workflow includes cell lysis reagents with gDNA removal, new Fast Advanced RT enzyme mix, buffer, and new TaqMan Fast Advanced Master Mix
  • Fast—7-minute sample prep, including DNase treatment, at room temperature
  • Easy—lyse samples in a tube or directly in culture plates
  • Robust—perform gene expression analysis on 10–100,000 cells per sample; results equivalent to those from purified RNA
  • Efficient—contains sufficient reagents to generate 500 real-time PCR results from 100 starting samples

Unique method eliminates the need to isolate RNA from cultured cells

The Cells-to-Ct method features a unique lysis reagent that lyses cells and protects RNA. DNA removal occurs simultaneously in the 5-minute lysis step to save time and improve ease of use. Lysis is then stopped by the addition of a 'Stop Solution' that irreversibly binds to the lysis reagents.

The TaqMan Fast Advanced Cells-to-CT Kit contains reverse transcription (RT) reagents for cDNA synthesis and TaqMan Fast Advanced Master Mix for real-time PCR analysis. TaqMan primer/probe sets are sold separately.

Simple 7-minute sample preparation

The TaqMan Fast Advanced Cells-to-CT Kit is designed for 10–100,000 cultured cells/sample. Cells are washed in PBS and lysed in solution for five minutes at room temperature; DNase treatment can be performed simultaneously. Lysis is terminated at room temperature by a 2-minute incubation with Stop Solution. The lysates are now ready for reverse transcription or storage at -20°C for a short term or -80°C for a long term.

Maximize sensitivity of detection

Unlike some competitor kits that limit the amount of lysate in the RT reaction to 10%, the TaqMan Fast Advanced Cells-to-CT Kit can accommodate 45% of the total RT reaction volume as cell lysate. Additionally, cDNA can comprise up to 45% of the real-time PCR reaction volume. The large lysate volume in the optimized RT reaction, along with the large cDNA volume in the subsequent real-time PCR using the TaqMan Fast Advanced Master Mix, lead to maximum sensitivity. The master mix amplifies the target precisely and accurately, enabling the detection of small quantities of target, such as transcripts expressed at low levels.

Reduced sample loss and transfer error

Because samples can be processed directly in culture wells (96- or 384-wells), sample handling and the potential for sample loss or transfer error are minimized, facilitating rapid, high-throughput processing. Unlike old-fashioned, multi-step RNA isolation protocols, no heating, washing, or centrifugation steps are required. The kit greatly simplifies a laborious 30–60 minute process and reduces it to 7 minutes.

Compatible with gold standard TaqMan Gene Expression Assays

The TaqMan Fast Advanced Cells-to-CT Kit workflow enables unsurpassed gene expression evaluation with any of the >700,000 TaqMan Gene Expression Assays. This kit has been extensively tested for specificity with a broad selection of TaqMan Gene Expression Assays.

Proven performance, proven together

All components of the TaqMan Fast Advanced Cells-to-CT Kit have been optimized for consistent and reliable performance. This removes the guesswork involved in assembling separate sample preparation, RT, and real-time PCR kits. And the TaqMan Fast Advanced Cells-to-CT Kit has been validated with TaqMan Gene Expression Assays for added quality assurance, giving you the sensitivity you need in the fastest sample-to-answer workflow available.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 20X Fast Advanced RT Enzyme Mix, 250 μL
• 2X Fast Advanced RT Buffer, 5 mL
• Lysis Solution, 5 mL
• Stop Solution, 550 μL
• DNase I, 55 μL
• TaqMan Fast Advanced Master Mix, 5 mL

Store at -5°C to -30°C.
Tube
2-step RT-qPCR
55 μL DNase I
High-throughput Compatible
Fast Advanced Cells-to-Ct Kit
100 Reactions
2X
100 Reactions
Cell Lysis
10 to 10,000 cells
TaqMan
High
7 min.
Fewer resources used, less waste, less hazardous
Cells-to-CT™
Dry Ice
Gene Expression
2X
Lysate
Mammalian cells
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.