missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TaqMan™ TaqMan™ Genotyping Master Mix
Optimized for end-point fluorescence detection in SNP genotyping applications.
139.00 € - 9120.00 €
Specifications
Detection Method | Primer-probe |
---|---|
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | AmpliTaq Gold DNA Polymerase |
Reaction Speed | Standard |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10390065
|
TaqMan™
4371353 |
1 x 1 mL |
139.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10025894
|
TaqMan™
4371355 |
1 x 10 mL |
1094.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10610606
|
TaqMan™
4381656 |
2 x 10 mL |
1998.00 €
20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10742877
|
TaqMan™
4371357 |
1 x 50 mL |
4650.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10239534
|
TaqMan™
4381657 |
2 x 50 mL |
9120.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Distinct clusters and high call rates for unambiguous allelic discrimination
- Validated with TaqMan™ SNP Genotyping Assays
- Excellent pre- and post-PCR stability for high throughput setup and analysis
- Reaction size is based on a 50μL reaction volume
May be used for a variety of genotyping applications, including:
- Candidate gene studies
- Drug-target validation
- Disease association studies
- Population genetics
- Linkage mapping
- Agricultural applications
Exceptional Performance from an Optimized Mix (2X)
Accurate genotype assignments result from preferential binding of the allele-specific probe to the matching target. TaqMan™ Genotyping Master Mix enables specific binding of the probe to achieve exceptional cluster resolution. Key features include:
- AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure) - Automatic hot start enzyme designed to be active during thermal cycling and inactive at room temperature for easy reaction setup
- Optimized mix components provide excellent specificity for discrimination between alleles
- Passive internal reference based on proprietary ROX™ dye for increased precision on Applied Biosystems™ real-time PCR instruments
- Single thermal cycling condition for consistent results with TaqMan™ Assays
Validated with TaqMan™ Genotyping Assays and Instruments:
- TaqMan™ Genotyping Master Mix has been tested across all types of TaqMan™ SNP Genotyping Assays: TaqMan™ Drug Metabolism Genotyping Assays, TaqMan™ SNP Genotyping Assays, and Custom TaqMan™ SNP Genotyping Assays
- In addition, the master mix is validated with the Applied Biosystems™ thermal cyclers and real-time PCR systems
High-Throughput Setup and Analysis:
- The combination of several low-cost thermal cyclers for PCR and a single real-time PCR instrument for allelic discrimination make high throughput SNP genotyping manageable
- TaqMan™ Genotyping Master Mix further improves high-throughput setup and analysis with excellent room temperature stability both before and after PCR
- This provides the flexibility required to run long experiments, unattended overnight or over a weekend
Reliable Discrimination with Challenging Targets
- Amplicon design constraints make SNP detection challenging, particularly when targets contain an abundance of GC- or AT-rich regions
- The optimized components of TaqMan™ Genotyping Master Mix result in tight, well-separated clusters and more accurate allele-calls compared to other mixes, even for challenging targets
Setting the Industry Standard for Quality:
- Extensive analytical and functional tests are performed for each manufactured lot of TaqMan™ Genotyping Master Mix, including one TaqMan™ SNP Genotyping Assay and one TaqMan™ Drug Metabolism Genotyping Assay
- Results are compiled in an informative Certificate of Analysis, which is accessible through the Applied Biosystems™ Web site
Order Info
Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice
Specifications
Primer-probe | |
qPCR | |
Standard | |
Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 | |
ROX (Pre-mixed) | |
Genotyping Master Mix | |
Genotyping | |
2 X |
High | |
AmpliTaq Gold DNA Polymerase | |
SNP Genotyping (Real-Time PCR-Based), CNV (Copy Number Variation) | |
Copy Number Variants, SNPs (Unknown or Numerous), Drug Metabolizing Enzymes, SNPs (Known) | |
TaqMan™ | |
UP (Ultra Pure) | |
2X | |
DNA (Genomic) |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title