missing translation for 'onlineSavingsMsg'
Learn More

TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™

23.80 € - 54.90 €

Specifications

Chemical Name or Material Tris Buffered Saline
Name Note 10X Solution
CAS 7447-40-7, 7647-01-0, 7647-14-5, 77-86-1, 7732-18-5
Boiling Point 100°C
Color Colorless
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10648973
View Documents
Fisher BioReagents
BP2471-100
100 mL Poly Bottle
23.80 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10153103
View Documents
Fisher BioReagents
BP2471-500
500 mL Poly Bottle
42.10 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10776834
View Documents
Fisher BioReagents
BP2471-1
1 L Nalgene Poly Bottle
54.90 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology

Washing buffer for alkaline phosphatase or peroxidase conjugates in Western blotting
Specifications

Specifications

Tris Buffered Saline
7447-40-7, 7647-01-0, 7647-14-5, 77-86-1, 7732-18-5
Colorless
Liquid
DNase-, RNase- and Protease-Free
TBS
10X solution contains 1.37M Sodium Chloride, 0.027M Potassium Chloride, and 0.25M Tris/Tris-HCl.
10X Solution
100°C
7.5
DNase free
Protease free
Molecular Biology
Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.