Learn More
Tocris™ TC SL C5
Metastasis-associated lung adenocarcinoma transcript 1 (Malat1) RNA inhibitor
Brand:  Tocris™ 6884/50
390.49 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18745796
  • 494.00 € / 50mg
Explore more special offers
Description
Description
TC SL C5 is a metastasis-associated lung adenocarcinoma transcript 1 (Malat1) RNA inhibitor (KD = 2.9 μM). Binds Malat1 3' terminal stability element for nuclear expression (ENE) triplex. Reduces Malat1 RNA levels by 54% and branching morphogenesis by 38% in mammary tumor organoids. Modulates Malat1 downstream genes without affecting Neat1.
Specifications
Specifications
50 mg
323.4
98%
C19H21N3O2
Inhibitor