missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ TC SL C5
Metastasis-associated lung adenocarcinoma transcript 1 (Malat1) RNA inhibitor
139.00 € - 494.00 €
Specifications
Molecular Formula | C19H21N3O2 |
---|---|
Formula Weight | 323.4 |
Product Type | Inhibitor |
Purity | 98% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18759306
|
Tocris™
6884/10 |
10 mg |
139.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18745796
|
Tocris™
6884/50 |
50 mg |
494.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TC SL C5 is a metastasis-associated lung adenocarcinoma transcript 1 (Malat1) RNA inhibitor (KD = 2.9 μM). Binds Malat1 3' terminal stability element for nuclear expression (ENE) triplex. Reduces Malat1 RNA levels by 54% and branching morphogenesis by 38% in mammary tumor organoids. Modulates Malat1 downstream genes without affecting Neat1.Specifications
C19H21N3O2 | |
323.4 | |
Inhibitor | |
98% |
Safety and Handling
Recommended Storage : Store at -20°C