Learn More
Tocris™ TC SL C5
Metastasis-associated lung adenocarcinoma transcript 1 (Malat1) RNA inhibitor
Brand: Tocris™ 6884/10
108.86 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
TC SL C5 is a metastasis-associated lung adenocarcinoma transcript 1 (Malat1) RNA inhibitor (KD = 2.9 μM). Binds Malat1 3' terminal stability element for nuclear expression (ENE) triplex. Reduces Malat1 RNA levels by 54% and branching morphogenesis by 38% in mammary tumor organoids. Modulates Malat1 downstream genes without affecting Neat1.Specifications
10 mg | |
323.4 | |
98% |
C19H21N3O2 | |
Inhibitor |