missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Use directly to resuspend and/or dilute purified DNA or RNA
44.70 € - 70.35 €
Chemical Identifiers
CAS | 1185-53-1 |
---|---|
Molecular Formula | C4H12ClNO3 |
Molecular Weight (g/mol) | 157.594 |
InChI Key | QKNYBSVHEMOAJP-UHFFFAOYSA-N |
PubChem CID | 93573 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
SMILES | C(C(CO)(CO)N)O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15875228
|
Thermo Scientific
J75893.X^ |
10 x 1 mL | Nalgene bottle |
44.70 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15661509
|
Thermo Scientific
J75893.AE |
100 mL | Nalgene bottle |
66.80 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15875418
|
Thermo Scientific
J75893.AP |
500 mL | Nalgene bottle | ||||||
Description
Use directly to resuspend and/or dilute purified DNA or RNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1185-53-1 | |
157.594 | |
93573 | |
C(C(CO)(CO)N)O.Cl |
C4H12ClNO3 | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
Specifications
1185-53-1 | |
C4H12ClNO3 | |
C(C(CO)(CO)N)O.Cl | |
157.594 | |
Molecular Biology |
Solution | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
93573 | |
TE Buffer |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only