Learn More
TE Buffer, Tris-EDTA, 100X Solution, pH 8.0, Molecular Biology, Fisher BioReagents™
65.90 € - 222.14 €
Specifications
Chemical Name or Material | Tris-EDTA |
---|---|
Name Note | 100X Solution |
CAS | 77-86-1, 60-00-4, 7732-18-5 |
Color | Undesignated |
Physical Form | Liquid |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10264643
|
Fisher BioReagents
BP1338-1 |
1 L | Nalgene Poly Bottle |
65.90 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10143393
|
Fisher BioReagents
BP1338-4 |
4 L | PolyPac™ | N/A | ||||||
Description
TE Buffer, Tris-EDTA, 100X Solution, pH 8.0, Molecular Biology
Tris–EDTA (TE) is routinely used for suspending nucleic acid samples.Specifications
Tris-EDTA | |
77-86-1, 60-00-4, 7732-18-5 | |
Liquid | |
Filtered through a 0.2-micron filter. | |
Molecular Biology |
100X Solution | |
Undesignated | |
DNase free | |
Protease free |
Safety and Handling

WARNING!
Emergency Overview
Irritating to eyes and skin. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.
NFPA
Health:2
Flammability:0
Instability:0
Recommended Storage : RT
Your input is important to us. Please complete this form to provide feedback related to the content on this product.