Learn More
Technical pH Buffer Solutions, METTLER TOLEDO™
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Provides a reliable all-around pH buffer. Mettler Toledo™ Technical pH Buffer Solution are specified to two decimal places and are traceable to NIST standards. Color-coded solution comes in a handy 250mL plastic bottle with no-fuss labels.
Brand: Mettler Toledo 51350012
25.89 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
pH measurements are only as accurate as the buffer solutions used for calibration purposes. These technical-grade buffer solutions provide a reliable pH buffer for regular use with most applications. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)
- NIST traceable and specified to two decimal places
- Handy screw-top plastic bottles for easy handling
- Includes a test certificate and no-fuss label with a temperature table
- SDS and label contains information according to GHS (Globally Harmonized System)
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Colorless | |
pH Electrode | |
250 mL | |
Plastic, screw-cap bottle | |
Technical |
pH measurement, electrode calibration | |
11.00 | |
Mild odor | |
Yes | |
1–2.5% Diisopropylamine |
Safety and Handling
- Technical buffer solution pH 11.00
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Diisopropylamine
Your input is important to us. Please complete this form to provide feedback related to the content on this product.