Learn More
Technical pH Buffer Solutions, METTLER TOLEDO™
Provides a reliable all-around pH buffer. Mettler Toledo™ Technical pH Buffer Solution are specified to two decimal places and are traceable to NIST standards. Color-coded solution comes in a handy 250mL plastic bottle with no-fuss labels.
27.35 € - 27.65 €
Specifications
For Use With (Application) | pH measurement, electrode calibration |
---|---|
For Use With (Equipment) | pH Electrode |
Quantity | 250 mL |
Traceability to NIST | Yes |
Grade | Technical |
Product Code | Brand | Color | pH | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | pH | Price | Quantity & Availability | ||||
10383191
|
Mettler Toledo
51350008 |
Blue | 9.21 |
27.65 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10343291
|
Mettler Toledo
51350010 |
Colorless | 10.00 |
27.60 €
250mL |
In Stock
Log in to see stock. |
||||
10634091
|
Mettler Toledo
51350012 |
Colorless | 11.00 |
27.35 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10598281
|
Mettler Toledo
51350002 |
Colorless | 2.00 |
27.60 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10683701
|
Mettler Toledo
51350006 |
Green | 7.00 |
27.55 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10201841
|
Mettler Toledo
51350004 |
Red | 4.01 |
27.55 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
pH measurements are only as accurate as the buffer solutions used for calibration purposes. These technical-grade buffer solutions provide a reliable pH buffer for regular use with most applications. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)
- NIST traceable and specified to two decimal places
- Handy screw-top plastic bottles for easy handling
- Includes a test certificate and no-fuss label with a temperature table
- SDS and label contains information according to GHS (Globally Harmonized System)
Specifications
pH measurement, electrode calibration | |
250 mL | |
Technical |
pH Electrode | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.