Learn More
Memmert™ Temperature Restriction for UF75
Optional temperature restriction between 60°C and 250°C.
Brand:  Memmert™ UF75A8
Product Code. 17529635
  • 164.00 € / Each
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • 1 Choice
  • Temperatures: 60°C, 70°C, 80°C, 95°C, 100°C, 120°C, 160°C, 180°C, 200°C, or 250°C
Specifications
Specifications
UF75 Oven
Temperature Restriction
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Temperature Restriction for UF75 > For Use With: UF75