Learn More
Bochem™ Tempered Cast Iron Bossheads
Manufactured from malleable cast iron (DIN 12895). Bochem™ Tempered Cast Iron Bossheads feature thumb and safety screws.
Brand: Bochem™ 5361
This item is not returnable.
View return policy
Description
- Malleable cast iron (DIN 12895)
- Type 1: Thumb screw M8
- Type 4: Safety screw type "heart" M8
Specifications
Tempered Cast Iron | |
Tempered Cast Iron Bossheads | |
Screw Type: Cross |
16.5 mm | |
8 | |
1 Ea. |