missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Tempered Cast Iron Bossheads

Manufactured from malleable cast iron (DIN 12895). Bochem™ Tempered Cast Iron Bossheads feature thumb and safety screws.

12.60 € - 22.55 €

Specifications

Material Tempered Cast Iron
Product Type Tempered Cast Iron Bossheads
Thread Size 8
Quantity 1 Ea.
View More Specs

Products 4
Product Code Brand Diameter (Metric) Outer Description Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Description Price Quantity & Availability  
10120811
View Documents
Bochem™
5361
16.5 mm Screw Type: Cross
13.00 €
Each
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
 
10354171
View Documents
Bochem™
5360
16.8 mm Screw Type: 1 Thumb screw M8
12.60 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
 
10604701
View Documents
Bochem™
5362
16.5 mm Screw Type: Swivel
19.45 €
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
 
10271632
View Documents
Bochem™
5369
36 mm Screw Type: 1 Thumb screw M8
22.55 €
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
Description

Description

  • Malleable cast iron (DIN 12895)
  • Type 1: Thumb screw M8
  • Type 4: Safety screw type "heart" M8
Specifications

Specifications

Tempered Cast Iron
Tempered Cast Iron Bossheads
8
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.