Learn More
Bochem™ Tempered Cast Iron Bossheads
Manufactured from malleable cast iron (DIN 12895). Bochem™ Tempered Cast Iron Bossheads feature thumb and safety screws.
Brand:  Bochem™ 5360
 View more versions of this product
Product Code. 10354171
  • 12.60 € / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Malleable cast iron (DIN 12895)
  • Type 1: Thumb screw M8
  • Type 4: Safety screw type "heart" M8
Specifications
Specifications
Tempered Cast Iron
Tempered Cast Iron Bossheads
Screw Type: 1 Thumb screw M8
16.8 mm
8
1 Ea.