Learn More
tert-Butyl 2-aminoisobutyrate hydrochloride, 98%, Thermo Scientific Chemicals
56.00 € - 656.00 €
Chemical Identifiers
CAS | 84758-81-6 |
---|---|
Molecular Formula | C8H17NO2 |
Molecular Weight (g/mol) | 159.23 |
MDL Number | MFCD00237698 |
InChI Key | LSVYCJILORYVCD-UHFFFAOYSA-N |
Synonym | alpha-aminoisobutyric acid t-butyl ester, h-aib-otbu.hcl, tert-butyl 2-methylalaninate, 2-aminoisobutyric acid t-butyl ester, alanine, 2-methyl-, 1,1-dimethylethyl ester, 2-amino-2-methylpropionic acid tert-butyl ester, h-aib-otbu inverted exclamation mark currencyhcl, t-butyl 2-methylalaninate, 2-methylalanine t-butyl ester, h-alpha-me-ala-otbu . hcl |
PubChem CID | 7019940 |
IUPAC Name | tert-butyl 2-amino-2-methylpropanoate |
SMILES | CC(C)(C)OC(=O)C(C)(C)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478536
|
Thermo Scientific Alfa Aesar
H63817.MD |
250 mg | N/A | ||||||
15410719
|
Thermo Scientific Alfa Aesar
H63817.03 |
1 g |
168.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15420719
|
Thermo Scientific Alfa Aesar
H63817.06 |
5 g |
656.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
tert-Butyl 2-aminoisobutyrate hydrochloride is used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstert-Butyl 2-aminoisobutyrate hydrochloride is used as intermediates.
Solubility
Soluble in water or 1% acetic acid.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Chemical Identifiers
84758-81-6 | |
159.23 | |
LSVYCJILORYVCD-UHFFFAOYSA-N | |
7019940 | |
CC(C)(C)OC(=O)C(C)(C)N |
C8H17NO2 | |
MFCD00237698 | |
alpha-aminoisobutyric acid t-butyl ester, h-aib-otbu.hcl, tert-butyl 2-methylalaninate, 2-aminoisobutyric acid t-butyl ester, alanine, 2-methyl-, 1,1-dimethylethyl ester, 2-amino-2-methylpropionic acid tert-butyl ester, h-aib-otbu inverted exclamation mark currencyhcl, t-butyl 2-methylalaninate, 2-methylalanine t-butyl ester, h-alpha-me-ala-otbu . hcl | |
tert-butyl 2-amino-2-methylpropanoate |
Specifications
84758-81-6 | |
C8H17NO2 | |
alpha-aminoisobutyric acid t-butyl ester, h-aib-otbu.hcl, tert-butyl 2-methylalaninate, 2-aminoisobutyric acid t-butyl ester, alanine, 2-methyl-, 1,1-dimethylethyl ester, 2-amino-2-methylpropionic acid tert-butyl ester, h-aib-otbu inverted exclamation mark currencyhcl, t-butyl 2-methylalaninate, 2-methylalanine t-butyl ester, h-alpha-me-ala-otbu . hcl | |
LSVYCJILORYVCD-UHFFFAOYSA-N | |
tert-butyl 2-amino-2-methylpropanoate | |
7019940 | |
98% | |
tert-Butyl 2-aminoisobutyrate hydrochloride |
White | |
MFCD00237698 | |
Soluble in water or 1% acetic acid. | |
CC(C)(C)OC(=O)C(C)(C)N | |
159.23 | |
195.69 | |
Powder |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.