missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl methyl ether, 99.8%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals Product Code.: 10172964

tert-Butyl methyl ether, 99.8%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals

Product Code. 10172964
2.5 L, Glass Bottle
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10172964

Brand: Thermo Scientific Acros 428010025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -110.0°C
Absorbance 1 cm cell vs HPLC-grade water, 0.097 max. at 255 nm, 0.009 max. at 280 nm, 1 cm cell vs HPLC-grade water at 0 nm
CAS 1634-04-4
Color Colorless
Density 0.7404g/mL
Boiling Point 54.0°C to 56.0°C
Quantity 2.5 L
Flash Point −28°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3675 to 1.3695
Assay Percent Range 99.75% min. (GC)
Residue after Evaporation 0.0003% max.
Specific Gravity 0.7404
Molecular Formula C5H12O
Linear Formula (CH3)3COCH3
MDL Number MFCD00008812
UN Number 2398
Viscosity 0.36 mPa.s (20°C)
Water 0.02% max.
Merck Index 15, 6106
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
Solubility Information Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
SMILES CC(C)(C)OC
IUPAC Name 2-methoxy-2-methylpropane
Molecular Weight (g/mol) 88.15
PubChem CID 15413
ChEBI CHEBI:27642
Formula Weight 88.15
Percent Purity 99.8%
Grade Spectroscopy
Chemical Name or Material tert-Butyl methyl ether, For spectroscopy
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467042 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number428010025
Lot NumberA0453892 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number428010025
Lot NumberA0405216 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number428010025
Lot NumberA0417794 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number428010025
Lot NumberA0439586 Certificate TypeCertificate of Analysis Date02/03/2022 Catalog Number428010025
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl methyl ether, 99.8%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.